Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h2bc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290380.1 | NP_001277309.1 | 1 | 109 | ||
NM_023422.3 | NP_075911.2 | 1 | 109 | |||
GenBank mRNA | AK005191.1 | - | 1 | 104 | ||
AK005407.1 | - | 1 | 104 | |||
AK011516.1 | - | 1 | 111 | |||
BC019673.1 | - | 1 | 92 | 3993954 | ||
BC082774.1 | - | 1 | 92 |
Gene Symbol: Hist1h2be | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177653.1 | NP_001171124.1 | 2 | 269 | ||
NM_001290530.1 | NP_001277459.1 | 1 | 90 | |||
NM_178194.4 | NP_835501.1 | 2 | 295 | |||
GenBank mRNA | AK030546.1 | - | 2 | 297 | ||
AK049948.1 | - | 2 | 270 | |||
BC069889.1 | - | 1 | 83 | 4217814 |
Gene Symbol: Hist1h2bf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178195.2 | NP_835502.1 | 1 | 76 |
Gene Symbol: Hist1h2bg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178196.4 | NP_835503.1 | 1 | 109 | ||
GenBank mRNA | BC060304.1 | - | 1 | 136 | 4217587 |
Gene Symbol: Hist1h2bh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178197.2 | NP_835504.1 | 1 | 168 | ||
GenBank mRNA | AK158416.1 | - | 117 | |||
BC092138.1 | - | 83 | 30613720 | |||
BC132584.1 | - | 1 | 168 | 40130861 | ||
BC132586.1 | - | 1 | 168 | 40130863 |
Gene Symbol: Hist1h2bj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178198.2 | NP_835505.1 | 1 | 76 | ||
GenBank mRNA | BC115792.1 | - | 68 |
Gene Symbol: Hist1h2bk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175665.2 | NP_783596.1 | 1 | 142 | ||
GenBank mRNA | BC147143.1 | - | 1 | 142 | 9056184 | |
BC147144.1 | - | 1 | 142 | 9056197 |
Gene Symbol: Hist1h2bl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178199.2 | NP_835506.1 | 1 | 76 |
Gene Symbol: Hist1h2bn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178201.2 | NP_835508.1 | 1 | 76 |
Gene Symbol: Hist1h2bq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097979.2 | NP_001091448.2 | 1 | 104 |
Gene Symbol: Hist1h2br | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110555.1 | NP_001104025.1 | 1 | 104 | ||
GenBank mRNA | AK036389.1 | - | 1 | 105 | ||
BC011440.1 | - | 1 | 89 | 3989862 |