Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pced1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114541.1 | NP_001108013.1 | 6 | 859 | ||
NM_178762.4 | NP_848877.2 | 5 | 1446 | |||
GenBank mRNA | AK031469.1 | - | 2 | 2577 | ||
AK032354.1 | - | 5 | 1438 | |||
AK032762.1 | - | 5 | 1421 | |||
AK041939.1 | - | 1 | 2245 | |||
AK044590.1 | - | 2 | 2143 | |||
AK045248.1 | - | 4 | 1516 | |||
AK077745.1 | - | 1 | 2250 | |||
AK079521.1 | - | 2095 | ||||
AK081584.1 | - | 4 | 1168 | |||
AK085613.1 | - | 4 | 1506 | |||
AK195014.1 | - | 1 | 13 | |||
BC064802.1 | - | 6 | 905 | 30009464 |