Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itpripl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033380.3 | NP_001028552.1 | 1 | 2801 | ||
GenBank mRNA | AK036744.1 | - | 1 | 628 | ||
AK086993.1 | - | 1 | 252 | |||
AK134593.1 | - | 1 | 2749 | |||
AK137678.1 | - | 1 | 2803 | |||
AK149729.1 | - | 1 | 2715 | |||
BC150856.1 | - | 1 | 2452 | 9087767 | ||
BC150864.1 | - | 1 | 2452 | 9087775 |