Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D5Ertd577e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177187.4 | NP_796161.3 | 4 | 2372 | ||
GenBank mRNA | AK054417.1 | - | 4 | 2372 | ||
BC053383.1 | - | 3 | 2281 | |||
BC094905.1 | - | 3 | 2269 | 30047197 | ||
BC099941.1 | - | 4 | 2356 | 30031929 |
Gene Symbol: Gm16513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200055.1 | NP_001186984.1 | 2416 |