Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001718.4 | NP_001709.1 | 5 | 1395 | ||
GenBank mRNA | AK300628.1 | - | 6 | 1355 | ||
BC096084.2 | - | 6 | 963 | |||
BC096085.2 | - | 6 | 963 | |||
BC099624.1 | - | 6 | 963 | |||
BC099625.1 | - | 6 | 1127 | |||
BC111763.1 | - | 6 | 1127 | |||
BC112352.1 | - | 6 | 1127 | |||
BC113018.1 | - | 6 | 1022 | |||
BC113019.1 | - | 6 | 1019 | |||
BC128031.1 | - | 6 | 919 | |||
M60315.1 | - | 5 | 1376 |