Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271463.1 | NP_001258392.1 | 21 | 2371 | ||
NM_001271464.1 | NP_001258393.1 | 20 | 2290 | |||
NM_001271465.1 | NP_001258394.1 | 21 | 2398 | |||
NM_177266.5 | NP_796240.3 | 21 | 2404 | |||
GenBank mRNA | AK035819.1 | - | 21 | 2309 | ||
AK044371.1 | - | 20 | 2291 | |||
AK052633.1 | - | 5 | 714 | |||
AK083441.1 | - | 20 | 2410 | |||
BC027341.1 | - | 21 | 2355 | 3984129 | ||
BC057062.1 | - | 10 | 2182 | |||
BC138233.1 | - | 21 | 2302 | 8861252 | ||
BC145208.1 | - | 21 | 2302 | 9053759 |