Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptrh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178595.3 | NP_848710.1 | 5 | 559 | ||
GenBank mRNA | AK075833.1 | - | 5 | 560 | ||
BC098218.1 | - | 5 | 599 | 3466952 |