Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ypel5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027166.5 | NP_081442.1 | 2 | 193 | ||
GenBank mRNA | AB098744.1 | - | 1 | 81 | ||
AK010201.1 | - | 2 | 159 | |||
AK046281.1 | - | 2 | 192 | |||
AK049096.1 | - | 2 | 194 | |||
AK049644.1 | - | 2 | 192 | |||
AK137446.1 | - | 2 | 165 | |||
AK154461.1 | - | 2 | 232 | |||
AK165943.1 | - | 2 | 195 | |||
AK167998.1 | - | 2 | 209 | |||
AK169241.1 | - | 2 | 196 | |||
AK169986.1 | - | 2 | 167 | |||
AK171010.1 | - | 2 | 194 | |||
AK217637.1 | - | 1 | 107 | |||
BC085109.1 | - | 2 | 212 | 30737658 |