Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001719.2 | NP_001710.1 | 6 | 1633 | ||
GenBank mRNA | AK094784.1 | - | 2 | 373 | ||
AK291186.1 | - | 6 | 1209 | |||
AK312419.1 | - | 6 | 1206 | |||
BC008584.1 | - | 6 | 1145 | 4183402 | ||
EU679366.1 | - | 6 | 1017 | |||
KJ875974.1 | - | 3 | 228 | |||
M60316.1 | - | 6 | 1200 | |||
X51801.1 | - | 6 | 1226 |