Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271665.1 | NP_001258594.1 | 4 | 729 | ||
NM_014886.4 | NP_055701.1 | 4 | 729 | |||
NR_073403.1 | 4 | 729 | ||||
XM_011543098.1 | XP_011541400.1 | 4 | 407 | |||
XM_017008952.1 | XP_016864441.1 | 4 | 407 | |||
XM_017008953.1 | XP_016864442.1 | 4 | 479 | |||
GenBank mRNA | AF077615.1 | - | 4 | 443 | ||
AF372458.1 | - | 4 | 454 | |||
AJ012409.1 | - | 4 | 441 | |||
AK313784.1 | - | 4 | 472 | |||
AY443419.1 | - | 3 | 359 | |||
BC005288.1 | - | 4 | 472 | 3683720 | ||
CB052084.1 | - | 2 | 94 |