Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134216.1 | NP_598977.1 | 1 | 881 | ||
GenBank mRNA | BC132625.1 | - | 1 | 981 | 40130902 | |
BC138067.1 | - | 1 | 981 | 8861086 |
Gene Symbol: Vmn1r207-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166709.1 | NP_001160181.1 | 1 | 881 |
Gene Symbol: Vmn1r209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013787.1 | NP_001013809.1 | 1 | 881 |