Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001719.2 | NP_001710.1 | 2 | 993 | ||
GenBank mRNA | AK291186.1 | - | 2 | 569 | ||
AK312419.1 | - | 2 | 566 | |||
BC008584.1 | - | 2 | 505 | 4183402 | ||
EU679366.1 | - | 2 | 377 | |||
M60316.1 | - | 2 | 560 | |||
X51801.1 | - | 2 | 586 |