Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128325.2 | NP_001121797.1 | 4 | 505 | ||
NM_001199021.1 | NP_001185950.1 | 5 | 859 | |||
NM_012445.3 | NP_036577.1 | 3 | 563 | |||
GenBank mRNA | AB027466.1 | - | 3 | 509 | ||
AK074618.1 | - | 3 | 370 | |||
AK074770.1 | - | 3 | 501 | |||
AK130164.1 | - | 3 | 281 | |||
AY358948.1 | - | 3 | 508 | |||
BC002707.2 | - | 3 | 293 | 3630467 | ||
BC036341.1 | - | 3 | 502 | 5183118 | ||
BY797606.2 | - | 3 | 547 |