Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001720.3 | NP_001711.2 | 7 | 2265 | ||
XM_011542024.2 | XP_011540326.2 | 8 | 2058 | |||
GenBank mRNA | AB075489.1 | - | 1 | 305 | ||
AY303955.1 | - | 7 | 2265 | |||
BC023526.2 | - | 7 | 1864 |