Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015795.1 | NP_056610.1 | 5 | 531 | ||
GenBank mRNA | AF176531.1 | - | 5 | 531 | ||
AK043554.1 | - | 5 | 501 | |||
AK081947.1 | - | 5 | 578 | |||
AK164657.1 | - | 5 | 614 | |||
BC086690.1 | - | 5 | 495 | 30683387 | ||
BC115890.1 | - | 5 | 489 | 40103787 |