Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppap2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005452.1 | - | 3 | 362 | ||
AK049581.1 | - | 3 | 405 | |||
AK177565.1 | - | 2 | 111 | |||
AK179023.1 | - | 2 | 166 | |||
AK208995.1 | - | 2 | 111 | |||
AK214880.1 | - | 2 | 135 | |||
BC010332.1 | - | 3 | 383 | 2654393 | ||
BC016635.1 | - | 3 | 333 | |||
BC085104.1 | - | 3 | 321 | |||
BQ571983.1 | - | 2 | 208 | |||
CA540403.1 | - | 3 | 328 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF123611.1 | - | 397 | Chr. - - [-] on Build |