Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026790.2 | NP_081066.2 | 7 | 972 | ||
NM_194066.2 | NP_919042.2 | 6 | 942 | |||
NM_194067.2 | NP_919043.1 | 6 | 837 | |||
NM_194068.2 | NP_919044.1 | 5 | 807 | |||
NM_194069.2 | NP_919045.1 | 4 | 687 | |||
GenBank mRNA | AJ566111.1 | - | 7 | 950 | ||
AJ566112.1 | - | 6 | 920 | |||
AJ566113.1 | - | 6 | 815 | |||
AJ566114.1 | - | 5 | 785 | |||
AJ566115.1 | - | 4 | 665 | |||
AK003665.1 | - | 6 | 776 | |||
AK075797.2 | - | 7 | 911 | |||
AK146231.1 | - | 6 | 814 | |||
AK169926.1 | - | 7 | 902 | |||
AK200168.1 | - | 2 | 324 | |||
BC125301.1 | - | 4 | 619 | 40129816 | ||
BC125636.1 | - | 4 | 619 | 40130151 | ||
BC128276.1 | - | 7 | 871 | 40091489 | ||
BY443016.1 | - | 2 | 344 |