Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vamp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016796.3 | NP_058076.2 | 4 | 247 | ||
GenBank mRNA | AF061516.1 | - | 3 | 219 | ||
AK002977.1 | - | 4 | 249 | |||
AK011678.1 | - | 4 | 227 | |||
AK018344.1 | - | 4 | 258 | |||
AK031151.1 | - | 4 | 249 | |||
BC138753.1 | - | 3 | 202 | 8861775 | ||
BC138754.1 | - | 3 | 202 | 8861776 |