Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164177.1 | NP_001157649.1 | 5 | 563 | ||
NM_016883.4 | NP_058579.2 | 5 | 642 | |||
GenBank mRNA | AB022022.1 | - | 5 | 598 | ||
AK009068.1 | - | 5 | 621 | |||
AK018233.1 | - | 5 | 620 | |||
AK136400.1 | - | 5 | 621 | |||
BC026931.1 | - | 5 | 609 | 3987112 | ||
BC056196.1 | - | 5 | 627 | 30019985 | ||
CD564300.1 | - | 5 | 530 |