Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001046637.1 | NP_001040102.1 | 6 | 1252 | ||
NM_001285825.1 | NP_001272754.1 | 7 | 1326 | |||
NM_015782.3 | NP_056597.3 | 7 | 1329 | |||
GenBank mRNA | AK013886.1 | - | 7 | 1303 | ||
AK088550.1 | - | 7 | 1185 | |||
AK210370.1 | - | 2 | 358 | |||
BC003229.1 | - | 7 | 1229 | 3483391 | ||
BC094006.1 | - | 7 | 1189 | 4983854 | ||
BC096648.1 | - | 7 | 1183 | 1397390 | ||
L15447.1 | - | 7 | 1125 |