Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vamp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080742.2 | NP_001074211.1 | 4 | 969 | ||
NM_016872.4 | NP_058568.1 | 3 | 1079 | |||
GenBank mRNA | AF119384.1 | - | 1 | 26 | ||
BC125557.1 | - | 1 | 16 | |||
BC125559.1 | - | 1 | 16 | |||
BC145146.1 | - | 1 | 16 | 9053697 |