Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017470.2 | NP_059498.2 | 2 | 288 | ||
GenBank mRNA | AB010031.1 | - | 2 | 266 | ||
AK002657.1 | - | 2 | 239 | |||
AK029618.1 | - | 1 | 99 | |||
AK132061.1 | - | 2 | 278 | |||
BC005426.1 | - | 2 | 217 | 3593932 | ||
BY012683.1 | - | 2 | 289 |