Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rragc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017475.2 | NP_059503.2 | 6, 7 | 1148 | ||
GenBank mRNA | AB017616.1 | - | 6, 7 | 1071 | ||
AK150792.1 | - | 6, 7 | 1103 | |||
AK159355.1 | - | 6, 7 | 1081 | |||
AK159434.1 | - | 6, 7 | 1103 | |||
AK166658.1 | - | 6, 7 | 1094 | |||
BC005417.1 | - | 6, 7 | 1096 | 3495022 | ||
BC037732.1 | - | 3, 4 | 445 | |||
BC071245.1 | - | 6, 7 | 1496 |