Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018740.2 | NP_061210.2 | 5, 6 | 733 | ||
GenBank mRNA | AF004107.1 | - | 4, 5 | 652 | ||
AK011130.1 | - | 3, 4 | 545 | |||
AK013662.1 | - | 3, 4 | 334 | |||
AK152582.1 | - | 5, 6 | 737 | |||
AK199980.1 | - | 2, 3 | 279 | |||
AK207946.1 | - | 1, 2 | 215 | |||
BC003752.1 | - | 4, 5 | 584 | 3489878 |