Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136260.1 | NP_001129732.1 | 19, 20 | 2696 | ||
NM_001197147.1 | NP_001184076.1 | 19, 20 | 2669 | |||
NM_018760.2 | NP_061230.2 | 19, 20 | 2696 | |||
GenBank mRNA | BC026592.1 | - | 10, 11 | 1489 | ||
BC129863.1 | - | 19, 20 | 2658 | |||
BC129864.1 | - | 19, 20 | 2657 | |||
BC148292.2 | - | 19, 20 | 2632 | 40110074 | ||
CB524421.1 | - | 4, 5 | 557 | |||
HQ018820.1 | - | 16, 17 | 2533 | |||
HQ204220.1 | - | 19, 20 | 2640 | |||
HQ285250.1 | - | 16, 17 | 2509 |