Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285870.1 | NP_001272799.1 | 6 | 1035 | ||
NM_001285871.1 | NP_001272800.1 | 6 | 1220 | |||
NM_001285873.1 | NP_001272802.1 | 6 | 1035 | |||
NM_018880.3 | NP_061368.1 | 6 | 1009 | |||
GenBank mRNA | AB030912.1 | - | 6 | 1036 | ||
AF220019.1 | - | 6 | 928 | |||
AK019165.1 | - | 6 | 970 | |||
AK144674.1 | - | 6 | 938 | |||
AK170029.1 | - | 6 | 973 | |||
BC003347.1 | - | 6 | 1050 | |||
BC034263.1 | - | 6 | 974 | 4503884 |