Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bgrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019989.2 | NP_064373.1 | 3 | 449 | ||
GenBank mRNA | AJ272504.1 | - | 3 | 319 | ||
AK049929.1 | - | 3 | 341 | |||
AK146588.1 | - | 3 | 357 | |||
AK150604.1 | - | 3 | 357 | |||
AK151095.1 | - | 3 | 359 | |||
AK151501.1 | - | 3 | 319 | |||
AK151871.1 | - | 3 | 361 | |||
AK165995.1 | - | 3 | 359 | |||
BC034161.1 | - | 3 | 403 | 4954986 | ||
BC043055.1 | - | 3 | 449 | 6418767 |