Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scamp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301634.1 | NP_001288563.1 | 4 | 545 | ||
NM_020270.3 | NP_064666.1 | 4 | 428 | |||
GenBank mRNA | AF241833.1 | - | 4 | 514 | ||
BC018613.1 | - | 4 | 392 | 4511417 |