Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320049.1 | NP_001306978.1 | 3 | 467 | ||
NM_006331.7 | NP_006322.4 | 3 | 467 | |||
NR_135131.1 | 3 | 467 | ||||
GenBank mRNA | AA314580.1 | - | 3 | 315 | ||
AK291597.1 | - | 3 | 332 | |||
AK298207.1 | - | 3 | 335 | |||
AK298745.1 | - | 3 | 327 | |||
BC055314.1 | - | 3 | 359 | 4718702 | ||
BM709304.1 | - | 3 | 396 | |||
BP315329.1 | - | 3 | 335 | |||
CD704778.1 | - | 3 | 456 | |||
DA070200.1 | - | 3 | 417 | |||
DC403138.1 | - | 3 | 467 | |||
U56427.1 | - | 3 | 225 | |||
U72514.1 | - | 3 | 310 |