Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srd5a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020611.4 | NP_065636.2 | 2 | 384 | ||
GenBank mRNA | AI553072.1 | - | 2 | 386 | ||
AK040198.1 | - | 2 | 352 | |||
AK075635.1 | - | 2 | 380 | |||
AK085748.1 | - | 2 | 233 | |||
AK138681.1 | - | 2 | 376 | |||
AK139290.1 | - | 2 | 381 | |||
AK162527.1 | - | 2 | 352 | |||
AK183560.1 | - | 1 | 90 | |||
BC039564.1 | - | 2 | 337 |