Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMPR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001203.2 | NP_001194.1 | 11 | 1369 | ||
NM_001256792.1 | NP_001243721.1 | 9 | 1265 | |||
NM_001256793.1 | NP_001243722.1 | 9 | 1206 | |||
NM_001256794.1 | NP_001243723.1 | 9 | 1249 | |||
XM_011532201.2 | XP_011530503.1 | 10 | 1248 | |||
XM_017008558.1 | XP_016864047.1 | 11 | 1440 | |||
XM_017008559.1 | XP_016864048.1 | 12 | 1430 | |||
XM_017008560.1 | XP_016864049.1 | 9 | 1322 | |||
XM_017008561.1 | XP_016864050.1 | 10 | 1279 | |||
GenBank mRNA | AK293008.1 | - | 9 | 1249 | ||
AK299930.1 | - | 9 | 1206 | |||
AK313642.1 | - | 9 | 1265 | |||
BC047773.1 | - | 8 | 1165 | 6202951 | ||
BC069796.1 | - | 10 | 1281 | 7262484 | ||
BC069803.1 | - | 10 | 1281 | 7262496 | ||
D89675.1 | - | 8 | 1113 | |||
U89326.1 | - | 11 | 1368 |