Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021493.2 | NP_067468.2 | 6 | 1793 | ||
GenBank mRNA | BQ444174.1 | - | 4 | 441 | ||
CB525670.1 | - | 4 | 336 |