Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_078479.3 | NP_510964.1 | 2 | 223 | ||
GenBank mRNA | AB049956.1 | - | 3 | 248 | ||
AK002588.1 | - | 3 | 251 | |||
AK003462.1 | - | 3 | 244 | |||
AK003602.1 | - | 3 | 243 | |||
AK007662.1 | - | 3 | 252 | |||
AK007838.1 | - | 2 | 234 | |||
AK008265.1 | - | 3 | 266 | |||
AK008293.1 | - | 3 | 245 | |||
AK009049.1 | - | 3 | 244 | |||
AK009703.1 | - | 3 | 252 | |||
BC027546.1 | - | 2 | 227 | 1515797 | ||
BC125658.1 | - | 2 | 224 | 40130173 | ||
BC125660.1 | - | 2 | 224 | 40130175 |