Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006336.3 | NP_006327.2 | 13 | 2336 | ||
XM_005251645.2 | XP_005251702.1 | 13 | 2128 | |||
XM_005251646.2 | XP_005251703.1 | 13 | 2325 | |||
XM_011518122.2 | XP_011516424.1 | 13 | 2138 | |||
XM_017014185.1 | XP_016869674.1 | 13 | 2744 | |||
XM_017014186.1 | XP_016869675.1 | 13 | 2119 | |||
XM_017014187.1 | XP_016869676.1 | 13 | 2738 | |||
XM_017014188.1 | XP_016869677.1 | 13 | 2106 | |||
GenBank mRNA | AK314911.1 | - | 13 | 2084 | ||
BC001447.2 | - | 2 | 131 | |||
BC052563.1 | - | 13 | 2321 | 6303965 | ||
X99802.1 | - | 12 | 1926 |