Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100502896 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277512.1 | NP_001264441.1 | 644 |
Gene Symbol: Gm11236 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277575.1 | NP_001264504.1 | 644 |
Gene Symbol: Gm11237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256481.2 | NP_001243410.2 | 5 | 644 |
Gene Symbol: Gm11487 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013393.1 | NP_001013411.1 | 5 | 728 | ||
GenBank mRNA | CK332043.1 | - | 2 | 194 |
Gene Symbol: Gm11757 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085538.2 | NP_001079007.1 | 698 | |||
GenBank mRNA | BG083055.2 | - | 551 |
Gene Symbol: Gm11758 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097978.2 | NP_001091447.2 | 730 | |||
GenBank mRNA | BC172045.1 | - | 5 | 698 | 9054331 |
Gene Symbol: Gm13871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177578.2 | NP_001171049.1 | 5 | 698 |
Gene Symbol: Gm428 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081644.1 | NP_001075113.1 | 730 | |||
GenBank mRNA | AK139501.1 | - | 732 |
Gene Symbol: LOC100862015 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277531.1 | NP_001264460.1 | 644 |
Gene Symbol: LOC101055863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277487.1 | NP_001264416.1 | 644 |