Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOMM40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128916.1 | NP_001122388.1 | 6, 7 | 756 | ||
NM_001128917.1 | NP_001122389.1 | 5, 6 | 848 | |||
NM_006114.2 | NP_006105.1 | 6, 7 | 753 | |||
XM_005258411.3 | XP_005258468.1 | 6, 7 | 750 | |||
GenBank mRNA | AF043250.1 | - | 6, 7 | 736 | ||
AF316401.1 | - | 5, 6 | 513 | |||
AF316402.1 | - | 5, 6 | 662 | |||
BC001779.1 | - | 5, 6 | 799 | 3354670 | ||
BC006413.1 | - | 6, 7 | 688 | 3609062 | ||
BC012134.1 | - | 6, 7 | 707 | 4549825 | ||
BC017224.2 | - | 5, 6 | 751 | 4298914 | ||
BC047528.1 | - | 6, 7 | 747 | 5297619 |