Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOMM40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128916.1 | NP_001122388.1 | 8 | 923 | ||
NM_001128917.1 | NP_001122389.1 | 7 | 1015 | |||
NM_006114.2 | NP_006105.1 | 8 | 920 | |||
XM_005258411.3 | XP_005258468.1 | 8 | 917 | |||
GenBank mRNA | AF043250.1 | - | 8 | 903 | ||
AF316398.1 | - | 6 | 519 | |||
AF316399.1 | - | 5 | 488 | |||
AF316401.1 | - | 7 | 680 | |||
AF316402.1 | - | 7 | 829 | |||
BC001779.1 | - | 7 | 966 | 3354670 | ||
BC006413.1 | - | 8 | 855 | 3609062 | ||
BC012134.1 | - | 8 | 874 | 4549825 | ||
BC017224.2 | - | 7 | 918 | 4298914 | ||
BC047528.1 | - | 8 | 914 | 5297619 |