Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarca5-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041999.1 | - | 1 | 949 | ||
AK079035.1 | - | 1 | 19 |
Gene Symbol: Gm13034 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK141203.1 | - | 1 | 1132 | ||
BC139113.1 | - | 1 | 1121 | 8862135 |