Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166010.1 | NP_001159482.1 | 2 | 219 | ||
NM_006117.2 | NP_006108.2 | 2 | 207 | |||
NM_206836.2 | NP_996667.2 | 2 | 217 | |||
NR_028588.1 | 2 | 207 | ||||
XM_006714957.1 | XP_006715020.1 | 2 | 210 | |||
XM_017010170.1 | XP_016865659.1 | 2 | 201 | |||
GenBank mRNA | AB209917.1 | - | 1 | 201 | ||
AF069301.1 | - | 2 | 167 | |||
AF153612.1 | - | 2 | 177 | |||
AF244138.1 | - | 2 | 172 | |||
AF257175.1 | - | 2 | 169 | |||
AK057175.1 | - | 2 | 210 | |||
AK075108.1 | - | 2 | 185 | |||
AK222617.1 | - | 2 | 193 | |||
AK223003.1 | - | 2 | 195 | |||
AK297049.1 | - | 2 | 185 | |||
AL136642.1 | - | 2 | 194 | |||
AU100345.1 | - | 2 | 207 | |||
AY297537.1 | - | 1 | 122 | |||
BC002668.2 | - | 2 | 194 | 3608151 | ||
BC016781.1 | - | 2 | 186 | 4107308 | ||
BC017474.1 | - | 2 | 171 | 3544703 | ||
BC025287.1 | - | 2 | 170 | |||
BC033841.1 | - | 2 | 177 | 3923244 | ||
BC034702.1 | - | 2 | 219 | 4750108 | ||
CR456768.1 | - | 1 | 90 |