Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018837.1 | NP_001018238.1 | 2 | 262 | ||
NM_006118.3 | NP_006109.2 | 2 | 406 | |||
GenBank mRNA | AK290626.1 | - | 2 | 328 | ||
AK294298.1 | - | 2 | 332 | |||
AK300676.1 | - | 2 | 360 | |||
AK310000.1 | - | 2 | 326 | |||
AK313291.1 | - | 2 | 337 | |||
BC005240.1 | - | 2 | 458 | 3931805 | ||
BC014314.1 | - | 2 | 327 | 3931852 | ||
BC015209.1 | - | 2 | 299 | 3883802 | ||
BC016730.1 | - | 2 | 344 | 4082810 | ||
BF668367.1 | - | 237 | ||||
EU190982.1 | - | 2 | 340 | |||
EU190983.1 | - | 2 | 323 | |||
KU178523.1 | - | 2 | 245 | |||
KU178524.1 | - | 2 | 101 | |||
U68566.1 | - | 2 | 406 |