Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAD2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127325.1 | NP_001120797.1 | 8 | 786 | ||
NM_006341.3 | NP_006332.3 | 8 | 753 | |||
XM_011540507.1 | XP_011538809.1 | 8 | 614 | |||
GenBank mRNA | AF072933.1 | - | 8 | 597 | ||
AF080398.1 | - | 7 | 582 | |||
AF139365.1 | - | 8 | 676 | |||
AF157482.1 | - | 8 | 753 | |||
AK027327.1 | - | 8 | 666 | |||
AK094316.1 | - | 10 | 1495 | |||
BC015244.1 | - | 7 | 567 | 3916742 |