Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MERTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006343.2 | NP_006334.2 | 11, 12 | 1811 | ||
XM_005263565.4 | XP_005263622.1 | 11, 12 | 1946 | |||
XM_005263568.4 | XP_005263625.1 | 11, 12 | 1946 | |||
XM_011510490.2 | XP_011508792.1 | 11, 12 | 1846 | |||
XM_017003164.1 | XP_016858653.1 | 11, 12 | 1816 | |||
XM_017003165.1 | XP_016858654.1 | 12, 13 | 1915 | |||
GenBank mRNA | AK294196.1 | - | 3, 4 | 257 | ||
AK308695.1 | - | 3, 4 | 259 | |||
AK316584.1 | - | 11, 12 | 1818 | |||
BC114917.1 | - | 11, 12 | 1525 | 40068588 | ||
BC114918.1 | - | 11, 12 | 1525 | 40012335 | ||
CN371300.1 | - | 1, 2 | 77 | |||
U08023.1 | - | 11, 12 | 1826 |