Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290230.1 | NP_001277159.1 | 3 | 628 | ||
NM_001798.4 | NP_001789.2 | 4 | 706 | |||
NM_052827.3 | NP_439892.2 | 4 | 706 | |||
XM_011537732.1 | XP_011536034.1 | 4 | 706 | |||
GenBank mRNA | AB012305.1 | - | 4 | 468 | ||
AK291941.1 | - | 4 | 645 | |||
AK293246.1 | - | 3 | 567 | |||
BC003065.2 | - | 4 | 626 | 3504348 | ||
BT006821.1 | - | 4 | 466 | |||
M68520.1 | - | 4 | 656 | |||
X61622.1 | - | 4 | 466 | |||
X62071.1 | - | 4 | 636 |