Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cndp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289531.1 | NP_001276460.1 | 7 | 1012 | ||
NM_023149.3 | NP_075638.2 | 8 | 980 | |||
GenBank mRNA | AB046738.1 | - | 8 | 921 | ||
AK003779.1 | - | 8 | 901 | |||
AK131124.1 | - | 7 | 892 | |||
AK149751.1 | - | 7 | 936 | |||
AK151013.1 | - | 8 | 902 | |||
AK152442.1 | - | 7 | 977 | |||
AK152729.1 | - | 7 | 950 | |||
AK153040.1 | - | 7 | 951 | |||
AK159129.1 | - | 7 | 979 | |||
AK159739.1 | - | 8 | 907 | |||
AK168029.1 | - | 8 | 919 | |||
AK168652.1 | - | 8 | 931 | |||
AK168856.1 | - | 6 | 669 | |||
AK172034.1 | - | 8 | 902 | |||
AK192792.1 | - | 3 | 231 | |||
BC005532.1 | - | 8 | 911 | 3496319 |