Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025349.2 | NP_079625.1 | 4 | 335 | ||
GenBank mRNA | AK003062.1 | - | 4 | 323 | ||
AK007462.1 | - | 4 | 332 | |||
AK009317.1 | - | 4 | 322 | |||
AK012446.1 | - | 4 | 332 | |||
AK013355.1 | - | 4 | 336 | |||
BC027511.1 | - | 4 | 325 | 1382565 | ||
BC081444.1 | - | 3 | 318 | 5711629 |