Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tomm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164729.1 | NP_001158201.1 | 3 | 827 | ||
NM_025365.3 | NP_079641.1 | 3 | 773 | |||
GenBank mRNA | AK003291.1 | - | 3 | 435 | ||
AK010470.1 | - | 3 | 485 | |||
AK019199.1 | - | 3 | 435 | |||
BC021590.1 | - | 2 | 959 | 5068609 | ||
BC037589.1 | - | 3 | 474 | 5007624 | ||
BC089325.1 | - | 1 | 1621 | |||
CA319049.1 | - | 3 | 476 | |||
CA321392.1 | - | 3 | 357 |