Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1110037F02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081183.1 | NP_001074652.1 | 6 | 591 | ||
GenBank mRNA | AK034232.1 | - | 6 | 513 | ||
AK052499.1 | - | 6 | 529 | |||
AK076731.1 | - | 6 | 561 | |||
AK122509.1 | - | 6 | 494 | |||
AK155623.1 | - | 6 | 592 | |||
AK162531.1 | - | 6 | 560 | |||
AK170075.1 | - | 6 | 594 |