Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ier3ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025409.3 | NP_079685.1 | 3 | 307 | ||
GenBank mRNA | AK004281.1 | - | 3 | 292 | ||
AK007388.1 | - | 3 | 255 | |||
AK007862.1 | - | 3 | 239 | |||
AK009116.1 | - | 3 | 240 | |||
AK190140.1 | - | 3 | 178 | |||
AK210106.1 | - | 3 | 175 | |||
BC021512.1 | - | 3 | 297 | 5351700 | ||
CF951253.1 | - | 2 | 38 | |||
CX207811.1 | - | 3 | 296 |