Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104530.1 | NP_001098000.1 | 3 | 890 | ||
NM_001104556.1 | NP_001098026.1 | 2 | 888 | |||
NM_001104557.1 | NP_001098027.1 | 2 | 720 | |||
NM_025426.3 | NP_079702.3 | 3 | 1058 | |||
GenBank mRNA | AK004354.1 | - | 3 | 1023 | ||
AK010357.1 | - | 3 | 886 | |||
AK012799.1 | - | 3 | 1038 | |||
AK144918.1 | - | 2 | 846 | |||
AK147739.1 | - | 3 | 1048 | |||
AK167407.1 | - | 3 | 1018 | |||
AK210054.1 | - | 1 | 21 | |||
BC011333.1 | - | 3 | 1045 | 3993084 | ||
CF983034.1 | - | 1 | 52 | |||
DV649959.1 | - | 1 | 27 |