Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037762.1 | NP_001032851.1 | 5 | 936 | ||
NM_025428.2 | NP_079704.2 | 5 | 978 | |||
GenBank mRNA | AK004475.1 | - | 5 | 879 | ||
AK013074.1 | - | 5 | 965 | |||
AK087676.1 | - | 5 | 950 | |||
BC021432.1 | - | 5 | 918 | 5030263 |